Merck
  • Bisphenol A release from orthodontic adhesives and its correlation with the degree of conversion.
Bisphenol A release from orthodontic adhesives and its correlation with the degree of conversion.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics (2014-12-24)
Deenadayalan Purushothaman, Vignesh Kailasam, Arun B Chitharanjan
ABSTRACT
Our objective was to quantitatively assess and compare the bisphenol A (BPA) released from an orthodontic adhesive using a light-emitting diode device (LED) or a halogen light-curing unit (HLC) at 3 tip-to-bracket distances (0, 5, and 10 mm) and varying curing times using high-performance liquid chromatography. BPA release with self-etching and moisture-insensitive primers with light-cured and chemically cured composites was also evaluated. BPA release was correlated to the corresponding degree of conversion. Our sample consisted of 598 stainless steel first premolar brackets. Of these, 520 were used for assessing BPA release and divided into 13 groups of 40 each. In groups I, II, and III, the composite was cured with the LED for 20 seconds at distances of 0, 5, and 10 mm, respectively. Groups IV, V, and VI were cured with the HLC for 40 seconds at the same 3 distances. Groups VII and VIII were cured for 5 and 10 seconds with the LED, and groups IX and X were cured for 10 and 20 seconds with the HLC at 0-mm distance. Groups XI, XII, and XIII consisted of brackets bonded with a self-etching primer and Transbond (3M Unitek, Monrovia, Calif), with a moisture-insensitive primer and Transbond, and with a chemically cured composite. The remaining 78 brackets were also divided into 13 groups and used for assessing the degree of conversion. The LED devices demonstrated significantly less BPA release and greater degrees of conversion (P <0.05). For both units, BPA release increased and the degree of conversion decreased as the tip distance increased and curing time decreased. The chemically cured group showed significantly less BPA release (P <0.05). Among the light-cured composites, those cured according to the manufacturers' recommendations (40 seconds and 0-mm distance for the HLC unit) released less BPA than did the self-etching primer and the moisture-insensitive primer. The degree of conversion was greatest for the chemically cured composite, whereas it was similar for the conventional, self-etching primer, and moisture-insensitive primer groups. However, correlations ranged from strongly negative to weakly positive between BPA release and degree of conversion. Clinicians should consider using LEDs in clinical practice and should keep the light-cure tip as close to the bracket as clinically possible. Curing time should be according to the manufacturer's recommendations. These steps will ensure less BPA release and a greater degree of conversion. Since chemically cured composites had less BPA release and a greater degree of conversion, they can be considered superior to light-cured composites in this aspect.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Bisphenol A, 97%
Supelco
Bisphenol A, ≥99%
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®