Merck
  • A practical guide for diagnosis and treatment of arteriovenous malformations in the oral and maxillofacial region.
A practical guide for diagnosis and treatment of arteriovenous malformations in the oral and maxillofacial region.
The Chinese journal of dental research : the official journal of the Scientific Section of the Chinese Stomatological Association (CSA) (2014-12-23)
Li Xin Su, Xin Dong Fan, Jia Wei Zheng, Yan An Wang, Zhong Ping Qin, Xu Kai Wang, Yao Wu Yang, Yi Fang Zhao
ABSTRACT
Arteriovenous malformations (AVMs) are congenital vascular malformations (CVMs) resulting from birth defects of the vasculature. It is rarely seen, only accounting for 1.5% of all vascular anomalies, and 50% of the lesions are located in the oral and maxillofacial region. Regardless of the type, AVMs may ultimately lead to significant anatomical, pathophysiological and hemodynamic consequences. Therefore, despite their relative rarity, AVMs still remain the most challenging and/or life-threatening form of vascular anomalies. Transarterial coil embolisation or ligation of feeding arteries are incorrect approaches and may result in progress of the lesion. Furthermore, such procedures would prevent future endovascular access to the lesions via the arterial route, and should be abandoned. Interventional embolisation using various sclerosants is currently the mainstay of treatment for AVMs, and elimination of the nidus (if present) is the key to success. Among various embolosclerotherapy agents, ethanol sclerotherapy produces the best long-term outcomes, with minimal complications. For more complex cases, multidisciplinary approaches and interventions may provide an excellent potential for a curative result. Based on the published literature and clinical experiences, a practical treatment guideline was established in order to provide a criterion for the management of oral and maxillofacial AVMs. This protocol will be renewed and updated to reflect cutting edge knowledge, and provide the newest treatment modalities for oral and maxillofacial AVMs.
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 8,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 150,000 (average), 99+% hydrolyzed
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol