Merck
  • Prior extended daily access to cocaine elevates the reward threshold in a conditioned place preference test.
Prior extended daily access to cocaine elevates the reward threshold in a conditioned place preference test.
Addiction biology (2013-05-03)
Zu-In Su, Jennifer Wenzel, Aaron Ettenberg, Osnat Ben-Shahar
ABSTRACT
We have previously shown that extended-access subjects exhibit heightened motivation for cocaine in the runway model, as reflected by reduced number of retreats. This heightened motivation could reflect either an increase in cocaine-induced reward or a decrease in cocaine-induced aversion. The current experiment was therefore devised to assess the cocaine-induced reward and aversion in extended-access rats using a place conditioning test. Rats trained to lever press for intravenous (IV) cocaine (0.25 mg/infusion) were provided 6-hour daily access to the drug over 10 days. Lever pressing in control subjects produced IV infusions of saline. Following drug self-administration, subjects underwent place conditioning for the immediate or delayed effects of cocaine (1.0 or 2.5 mg/kg, IV). In control subjects, the immediate effects of the low dose of cocaine produced conditioned places preferences (CPPs), while the delayed effects produced conditioned place aversions (CPAs). In contrast, the animals receiving low cocaine dose for 6 hours, exhibited place aversions but not preferences; an effect that was reversed when the dose of cocaine was increased. Additionally, in the 6-hour group, delayed conditioning was associated with a reduction in zif268 immunoreactivity in the medial prefrontal cortex and nucleus accumbens shell while immediate conditioning was associated with an increase in zif268-positive cells in the central nucleus of the amygdala. Collectively, these data suggest that extended daily access to cocaine produces a shift in the subject's perceived reward threshold that is paralleled by alterations in the activity of both the reward and stress pathways.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride hydrate, ≥96%
Sigma-Aldrich
Goat Serum Donor Herd, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)