Merck
  • Effect of a nutrient mixture on matrix metalloproteinase-9 dimers in various human cancer cell lines.
Effect of a nutrient mixture on matrix metalloproteinase-9 dimers in various human cancer cell lines.
International journal of oncology (2014-01-01)
M W Roomi, T Kalinovsky, M Rath, A Niedzwiecki
ABSTRACT
Strong clinical and experimental evidence demonstrates association of elevated levels of matrix metalloproteinase MMP-9 with cancer progression, metastasis and shortened patient survival, as it plays a key role in tumor cell invasion and metastasis by digesting the basement membrane and ECM components. MMP-9 is secreted in both the monomeric and dimeric form. Although there is little research on MMP-9 dimers, some studies have shown the dimer to be associated with more aggressive tumor progression. Our objective was to study the relative secretion patterns of MMP-9 monomer and dimer in a variety of cancer cell lines and the effect of a nutrient mixture (NM) containing lysine, proline, ascorbic acid and green tea extract on MMP-9 secretion. The cancer cell lines were grown in their respective media, supplemented with 10% FBS, penicillin (100 U/ml) and streptomycin (100 µg/ml) in 24-well tissue culture plates. At near confluence, the cells were treated with NM at 0,10, 50, 100, 500 and 1000 µg/ml. Parallel sets of cultures were treated with PMA (100 ng/ml) for induction of MMP-9. Cell MMP-9 secretion was assayed by gelatinase zymography. MMP-9 dimer secretion patterns of cancer cells fell into different categories. We observed no MMP-9 dimer in prostate DU-145 and PC-3, pancreatic MIA-Pa-Ca2, colon HCT-116, bladder T-24, head and neck FaDu, glioblastoma A-172, T-98 and LN-18 and leukemia HL-60, Jurkat, and Raji cell lines. MMP-dimer secretion only with PMA induction was seen in breast MCF-7 and MDA-MB-231, uterine SK-UT-1, lung A-549, tongue SC-25, melanoma A2058, osteosarcoma U-2OS, rhabdomyosarcoma, fibrosarcoma HT-1080, chondrosarcoma SW-1350 and liposarcoma SW-872. Cervical HeLa and DoTc 2 4510, renal 786-0 and HCC SK-Hep-1 cells exhibited MMP-9 dimer without PMA treatment and increased secretion with PMA treatment. Sarcomas had the highest levels of MMP-9 monomer and dimer with and without PMA among these cancer cell lines. Cervical, uterine and male breast cancer cell lines showed the next highest levels of MMP-9, followed by breast cancer cell lines. Melanoma, renal, lung, head and neck and HCC showed lower levels and prostate, glioblastoma, bladder and leukemia cell lines the lowest. NM showed dose-dependent inhibition of MMP-9 monomer and dimer in all cell lines tested. In conclusion, high MMP-9 and dimer secretion levels correlated with the most aggressive cancer cell lines. NM was effective in inhibiting MMP-9 and dimer secretion in all cell lines tested, suggesting its therapeutic potential as an antimetastatic agent.
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent
SAFC
L-Arginine
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, 99%
SAFC
L-Proline
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated