Merck
  • Lack of evidence for genomic instability in autistic children as measured by the cytokinesis-block micronucleus cytome assay.
Lack of evidence for genomic instability in autistic children as measured by the cytokinesis-block micronucleus cytome assay.
Autism research : official journal of the International Society for Autism Research (2014-11-06)
Penelope A E Main, Philip Thomas, Manya T Angley, Robyn Young, Adrian Esterman, Catherine E King, Michael F Fenech
ABSTRACT
Autism spectrum disorders are a set of neurodevelopmental disorders that are highly hereditable. Increased genomic instability has been observed in other heritable paediatric neurobiological disorders; therefore, the aim of our study was to test the hypothesis that DNA damage is increased in children with autism and that B vitamin status may explain variations in genome integrity between autistic and normal children. We compared 35 children with autism, 27 of their siblings without autism and 25 age- and gender-matched community controls for genomic stability using the cytokinesis-block micronucleus cytome (CBMN-cyt) assay, B vitamins and homocysteine, as well as autism-related behaviours. It was found that there were no differences in CBMN-cyt biomarkers between the three groups. Vitamin B2 was significantly raised in children with autism and their siblings compared with controls (P = 0.027 and P = 0.016 respectively) but there was no difference in other B vitamins or homocysteine. In conclusion, although replication using a larger cohort is needed, it appears unlikely that genomic instability is a feature of the aetiology of autism. We cannot rule out in utero effects or other types of DNA damage not measured by the CBMN-cyt assay.
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%