Merck
  • Simultaneous determination of diclofenac, its human metabolites and microbial nitration/nitrosation transformation products in wastewaters by liquid chromatography/quadrupole-linear ion trap mass spectrometry.
Simultaneous determination of diclofenac, its human metabolites and microbial nitration/nitrosation transformation products in wastewaters by liquid chromatography/quadrupole-linear ion trap mass spectrometry.
Journal of chromatography. A (2014-05-20)
Victoria Osorio, Marta Imbert-Bouchard, Bozo Zonja, José-Luis Abad, Sandra Pérez, Damià Barceló
ABSTRACT
An analytical method was developed and validated for the first determination of five major human metabolites of the non-steroidal anti-inflammatory drug diclofenac as well as two microbial transformation products in wastewater. The method was based on the extraction of diclofenac and the chemically synthetized compounds by solid-phase extraction (SPE), using a hydrophilic-lipophilic balanced polymer followed by liquid chromatography (LC) coupled to hybrid quadrupole-linear ion trap mass spectrometry (QqLIT-MS). Quantitation was performed by the internal standard approach, to correct for matrix effects. The accuracy of the method was generally higher than 40% for raw and treated wastewater with a precision below 12%. In wastewater influent and effluent samples the detection limits for the majority of target compounds were 0.3-2.5ngL(-1) and 0.1-3.1ngL(-1), respectively. The method was applied to the analysis of influent and effluent wastewater samples from urban wastewater treatment plants. Moreover, to obtain an extra tool for confirmation and identification of the studied diclofenac-derived compounds, Information-Dependent Acquisition (IDA) experiments were performed, with selected reaction monitoring (SRM) as the survey scan and an enhanced product ion (EPI) scan as the dependent scan. Diclofenac and its major human metabolite, 4'-hydroxydiclofenac were detected in all samples at concentrations of 331-1150ngL(-1) and 585-6000ngL(-1), respectively. Neither microbial transformation product of diclofenac was detected in any of the influent samples analyzed, but in effluents, their concentrations ranged from 4 to 105ngL(-1).
Product Description
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, analytical standard
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis