Merck
  • Beta-galactosidase staining in the nucleus of the solitary tract of Fos-Tau-LacZ mice is unaffected by monosodium glutamate taste stimulation.
Beta-galactosidase staining in the nucleus of the solitary tract of Fos-Tau-LacZ mice is unaffected by monosodium glutamate taste stimulation.
PloS one (2014-09-06)
Jennifer M Stratford, John A Thompson
ABSTRACT
Fos-Tau-LacZ (FTL) transgenic mice are used to visualize the anatomical connectivity of neurons that express c-Fos, an immediate early gene, in response to activation. In contrast to typical c-Fos protein expression, which is localized to the nucleus of stimulated neurons, activation of the c-Fos gene results in beta galactosidase (β-gal) expression throughout the entire cytoplasm of activated cells in FTL mice; thereby making it possible to discern the morphology of c-Fos expressing cells. This can be an especially important tool in brain areas in which function may be related to cell morphology, such as the primary taste/viscerosensory brainstem nucleus of the solitary tract (nTS). Thus, to further characterize FTL activity in the brain, the current study quantified both β-gal enzymatic activity as well as c-Fos protein expression in the nTS under a variety of experimental conditions (no stimulation, no stimulation with prior overnight food and water restriction, monosodium glutamate taste stimulation, and monosodium glutamate taste stimulation with perfusion 5 h post stimulation). Contrary to previous research, we found that β-gal activity (both labeled cell bodies and overall number of labeled pixels) was unchanged across all experimental conditions. However, traditional c-Fos protein activity (both cell bodies and number of activated pixels) varied significantly across experimental conditions, with the greatest amount of c-Fos protein label found in the group that received monosodium glutamate taste stimulation. Interestingly, although many c-Fos positive cells were also β-gal positive in the taste stimulated group, some c-Fos protein labeled cells were not co-labeled with β-gal. Together, these data suggest that β-gal staining within the nTS reflects a stable population of β-gal- positive neurons whose pattern of expression is unaffected by experimental condition.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture