Merck
  • Muscarinic M1 receptors regulate propofol modulation of GABAergic transmission in rat ventrolateral preoptic neurons.
Muscarinic M1 receptors regulate propofol modulation of GABAergic transmission in rat ventrolateral preoptic neurons.
Journal of molecular neuroscience : MN (2014-10-09)
Yu Zhang, Tian Yu, Yang Liu, Kun Qian, Bu-Wei Yu
ABSTRACT
GABAergic neurons within the ventrolateral preoptic area (VLPO) play an important role in sleep-wakefulness regulation. Propofol, a widely used systemic anesthetic, has lately been reported to excite noradrenaline (NA)-inhibited type of VLPO neurons. Present study tested if acetylcholine system takes part in the propofol modulation of GABAergic spontaneous miniature inhibitory postsynaptic currents (mIPSCs) in mechanically dissociated rat VLPO neurons using a conventional whole-cell patch clamp technique. Propofol reversibly decreased mIPSC frequency without affecting the current amplitude, indicating that propofol acts presynaptically to decrease the probability of spontaneous GABA release. The propofol action on GABAergic mIPSC frequency was completely blocked by atropine, a nonselective muscarinic acetylcholine (mACh) receptor antagonist, and pirenzepine, a selective M1 receptor antagonist. These results suggest that propofol acts on M1 receptors on GABAergic nerve terminals projecting to VLPO neurons to inhibit spontaneous GABA release. The M1 receptor-mediated modulation of GABAergic transmission onto VLPO neurons may contribute to the regulation of loss of consciousness induced by propofol.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Atropine, European Pharmacopoeia (EP) Reference Standard
Propofol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-DAMP, ≥98% (HPLC)