Merck
  • Immunological blockade of adipocyte inflammation caused by increased matrix metalloproteinase-cleaved osteopontin in obesity.
Immunological blockade of adipocyte inflammation caused by increased matrix metalloproteinase-cleaved osteopontin in obesity.
Obesity (Silver Spring, Md.) (2015-03-18)
Lukas Leitner, Karina Schuch, Alexander Jürets, Bianca K Itariu, Maike Keck, Viktor Grablowitz, Oskar C Aszmann, Gerhard Prager, Günther Staffler, Maximilian Zeyda, Thomas M Stulnig
ABSTRACT
Osteopontin (OPN) is upregulated in adipose tissue (AT) in obesity and contributes to subclinical inflammation, adipocyte dysfunction, and insulin resistance. OPN effects can be increased by cleavage by matrix metalloproteinases (MMP). This study aimed at investigating the presence of OPN cleavage products in human AT in obesity and their impact on adipocyte function and immunological blockade of these effects. AT of severely obese and control donors was investigated for OPN and MMP expression and the presence of OPN cleavage fragments. Primary adipocytes were isolated from human donors for in vitro investigation of cleaved OPN effects. OPN and MMP-9 expression was highly correlated in AT from obese donors, and increased levels of cleaved OPN were detected in AT from obese individuals. The in vitro effect of OPN on adipocyte inflammation and insulin resistance was enhanced by protease cleavage, which could finally be blocked with a monoclonal antibody directed against the MMP cleavage site of OPN. These findings show that MMP cleavage of OPN in AT occurs in obesity, thereby enhancing OPN's inflammatory and pro-diabetic activity on adipocytes. Specifically targeting MMP-cleaved OPN opens avenues for prevention and treatment of obesity-induced type 2 diabetes.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M