Merck
  • In vivo human skin penetration of (-)-epigallocatechin-3-gallate from topical formulations.
In vivo human skin penetration of (-)-epigallocatechin-3-gallate from topical formulations.
Acta pharmaceutica (Zagreb, Croatia) (2014-06-11)
Santo Scalia, Valentina Trotta, Anna Bianchi
ABSTRACT
The aim of the study was to examine the effect of topical vehicles on the in vivo human stratum corneum penetration of the antioxidant and skin photoprotective agent (-)-epigallocatechin-3-gallate (EGCG). Model oil-in-water (o/w) emulsion and gel formulations containing 1 % (m/m) EGCG were prepared and subjected to photodegradation studies in order to select excipients that minimize the light instability of EGCG. The optimized emulsion and gel were applied to human volunteers and the EGCG percutaneous permeation was evaluated in vivo by the tape- -stripping technique. No significant differences in the percentage of the applied EGCG dose diffused into the stratum corneum were observed between the o/w emulsion (36.1 ± 7.5 %) and gel (35.5 ± 8.1 %) preparations. However, the amount of EGCG permeated into the deeper region of human stratum corneum was significantly larger for the o/w emulsion compared to the gel. Therefore, the emulsion represents a suitable vehicle for topical delivery of EGCG.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Epigallocatechin gallate, primary reference standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
(−)-Epigallocatechin gallate, analytical standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%