Merck
  • Conjugation of a new series of dithiocarbazate Schiff base Copper(II) complexes with vectors selected to enhance antibacterial activity.
Conjugation of a new series of dithiocarbazate Schiff base Copper(II) complexes with vectors selected to enhance antibacterial activity.
Bioconjugate chemistry (2014-11-11)
May Lee Low, Laure Maigre, Pierre Dorlet, Régis Guillot, Jean-Marie Pagès, Karen A Crouse, Clotilde Policar, Nicolas Delsuc
ABSTRACT
A new series of six Schiff bases derived from S-methyldithiocarbazate (SMDTC) and S-benzyldithiocarbazate (SBDTC) with methyl levulinate (SMML, SBML), levulinic acid (SMLA, SBLA), and 4-carboxybenzaldehyde (SM4CB, SB4CB) were reacted with copper(II), producing complexes of general formula ML2 (M = Cu(II), L = ligand). All compounds were characterized using established physicochemical and spectroscopic methods. Crystal structures were determined for three Schiff bases (SMML, SBML, SBLA) and two Cu(II) complexes (Cu(SMML)2 and Cu(SMLA)2). In order to provide more insight into the behavior of the complexes in solution, electron paramagnetic resonance (EPR) and electrochemical experiments were performed. The parent ligands and their respective copper(II) complexes exhibited moderate antibacterial activity against both Gram-negative and Gram-positive bacteria. The most active ligand (SB4CB) and its analogous S-methyl derivative (SM4CB) were conjugated with various vector moieties: polyarginines (R1, R4, R9, and RW9), oligoethylene glycol (OEG), and an efflux pump blocker, phenylalanine-arginine-β-naphthylamide (PAβN). Nonaarginine (R9) derivatives showed the most encouraging synergistic effects upon conjugation and complexation with copper ion including enhanced water solubility, bacteria cell membrane permeability, and bioactivity. These Cu(II)-R9 derivatives display remarkable antibacterial activity against a wide spectrum of bacteria and, in particular, are highly efficacious against Staphylococcus aureus with minimum inhibitory concentration (MIC) values of 0.5-1 μM. This pioneer study clearly indicates that the conjugation of cell-penetrating peptides (CPPs) to dithiocarbazate compounds greatly enhances their therapeutic potential.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Polymyxin B nonapeptide hydrochloride, cationic cyclic peptide
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
HATU, ≥98.0% (CHN)
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Valeric acid, ≥99%, FCC, FG
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Supelco
Melting point standard 121-123°C, analytical standard