Merck
  • Effects of Galla chinensis extracts on UVB-irradiated MMP-1 production in hairless mice.
Effects of Galla chinensis extracts on UVB-irradiated MMP-1 production in hairless mice.
Journal of natural medicines (2014-09-18)
Zheng-wang Sun, Eunson Hwang, Hyun Ji Lee, Tae Youp Lee, Hyun Geun Song, Sang-Yong Park, Heon-Sub Shin, Don-Gil Lee, Tae Hoo Yi
ABSTRACT
Galla chinensis (GAC) is a natural traditional Chinese medicine that has been widely used in folk medicine. Although GAC compounds (mainly gallic acid and methyl gallate) possess strong antiviral, antibacterial, anticancer, and antioxidant activities, there is no report regarding topical or oral administration of GAC compounds on UVB irradiation-induced photoaging in hairless mice (SKH: HR-1). In the present study, we examined cell viability, intracellular reactive oxygen species (ROS), matrix metalloproteinase-1 (MMP-1), and interleukin-6 (IL-6) in skin fibroblasts and keratinocytes induced by UVB in vitro. We also studied skin damage by measuring skin thickness, elasticity, wrinkling and levels of protein MMP-1, elastin, procollagen type I, and transforming growth factor-β1 (TGF-β1) in hairless mouse skin chronically irradiated by UVB in vivo. GAC treatment significantly prevented skin photoaging by reducing the levels of ROS, MMP-1, and IL-6 and promoting production of elastin, procollagen type I, and TGF-β1. According to the results of H&E staining and Masson's trichrome staining, GAC reduced skin thickness and wrinkle formation while it increased skin elasticity. The effects of GAC on UVB-induced skin photoaging may be due to suppressed MMP-1 expression. These findings could be referenced for the development of new agents that target UVB-induced photoaging.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
USP
Retinyl palmitate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methyl 3,4,5-trihydroxybenzoate, 98%
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%