Merck
  • Sulforaphane alleviates muscular dystrophy in mdx mice by activation of Nrf2.
Sulforaphane alleviates muscular dystrophy in mdx mice by activation of Nrf2.
Journal of applied physiology (Bethesda, Md. : 1985) (2015-01-17)
Chengcao Sun, Cuili Yang, Ruilin Xue, Shujun Li, Ting Zhang, Lei Pan, Xuejiao Ma, Liang Wang, Dejia Li
ABSTRACT
Sulforaphane (SFN), one of the most important isothiocyanates in the human diet, is known to have chemo-preventive and antioxidant activities in different tissues via activation of nuclear factor erythroid 2-related factor 2 (Nrf2)-mediated induction of antioxidant/phase II enzymes, such as heme oxygenase-1 and NAD(P)H quinone oxidoreductase 1. However, its effects on muscular dystrophy remain unknown. This work was undertaken to evaluate the effects of SFN on Duchenne muscular dystrophy. Four-week-old mdx mice were treated with SFN by gavage (2 mg·kg body wt(-1)·day(-1) for 8 wk), and our results demonstrated that SFN treatment increased the expression and activity of muscle phase II enzymes NAD(P)H quinone oxidoreductase 1 and heme oxygenase-1 with a Nrf2-dependent manner. SFN significantly increased skeletal muscle mass, muscle force (∼30%), running distance (∼20%), and GSH-to-GSSG ratio (∼3.2-fold) of mdx mice and decreased the activities of plasma creatine phosphokinase (∼45%) and lactate dehydrogenase (∼40%), gastrocnemius hypertrophy (∼25%), myocardial hypertrophy (∼20%), and malondialdehyde levels (∼60%). Furthermore, SFN treatment also reduced the central nucleation (∼40%), fiber size variability, and inflammation and improved the sarcolemmal integrity of mdx mice. Collectively, these results show that SFN can improve muscle function and pathology and protect dystrophic muscle from oxidative damage in mdx mice associated with Nrf2 signaling pathway, which indicate Nrf2 may have clinical implications for the treatment of patients with muscular dystrophy.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
DL-Sulforaphane, ≥90% (HPLC), synthetic, liquid
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Menadione, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Menadione, United States Pharmacopeia (USP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard