Merck
  • Flavonoids inhibit COX-1 and COX-2 enzymes and cytokine/chemokine production in human whole blood.
Flavonoids inhibit COX-1 and COX-2 enzymes and cytokine/chemokine production in human whole blood.
Inflammation (2014-08-21)
Daniela Ribeiro, Marisa Freitas, Sara M Tomé, Artur M S Silva, Stefan Laufer, José L F C Lima, Eduarda Fernandes
ABSTRACT
Cyclooxygenase 2 (COX-2) and the production of cytokines/chemokines are important targets for the modulation of the inflammatory response. Although a large variety of inhibitors of these pathways have been commercialized, some of those inhibitors present severe side effects, governing the search for new molecules, as alternative anti-inflammatory agents. This study was undertaken to study an hitherto not evaluated group of flavonoids, concerning its capacity to inhibit COX-1 and COX-2 enzymes, as well as to inhibit the production of the cytokines and a chemokine, in a complex matrix involved in the systemic inflammatory process, the blood, aiming the establishment of a structure-activity relationship. The results obtained reveal promising flavonoids for the modulation of the inflammatory process, namely the ones presenting a catechol group in B ring, as some flavonoids were able to simultaneously inhibit the production of inflammatory prostaglandin E2 and pro-inflammatory cytokines.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
5Z-7-Oxozeaenol, ≥98% (HPLC)
Supelco
Apigenin, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acacetin, ≥97.0% (HPLC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
L-Glutamine
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
Taxifolin, analytical standard