Merck
  • Cytoplasmic TRAF4 contributes to the activation of p70s6k signaling pathway in breast cancer.
Cytoplasmic TRAF4 contributes to the activation of p70s6k signaling pathway in breast cancer.
Oncotarget (2015-03-05)
Hua-Yan Ren, Jian Wang, Fan Yang, Xiao-Li Zhang, Ai-Lian Wang, Li-Li Sun, Ke-Xin Diao, En-Hua Wang, Xiao-Yi Mi
ABSTRACT
Tumor necrosis factor receptor associated factor 4 (TRAF4) is an important adaptor protein that plays a significant role in several signaling pathways. By studying the relationship between TRAF4 and 70 kDa ribosomal protein S6 kinase (p70s6k) in vivo, we demonstrated that cytoplasmic TRAF4 was correlated with the activation of p70s6k in breast cancer. Moreover, we found that cytoplasmic TRAF4 expression in breast cancer patients was significantly associated with a poor prognosis. To determine the exact mechanism, we analyzed the interaction between TRAF4 and p70s6k and identified the Zinc fingers domain of TRAF4 was responsible for their interaction in MCF7 cells. Furthermore, we found that activation of p70s6k/S6 signaling pathway by TRAF4 requires the mammalian target of rapamycin (mTOR) activity; TRAF4 acted as a sensitizer. Tumor necrosis factor receptor associated factor 2 (TRAF2), as a binding partner of TRAF4, could also promoted activation of p70s6k signaling via upregulating cytoplasm expression of TRAF4 and played a critical role in TNFa-induced activation of p70s6k/S6 pathway. Finally, we demonstrated p70s6k/S6 signaling pathway played an important role in the promoting function of TRAF4 on cell proliferation. In summary, our work suggests a new direction for understanding the oncogenic function of TRAF4 in breast cancer.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Traf4
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
MISSION® esiRNA, targeting human TRAF4
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis