Merck
  • Over-expression of cofilin-1 suppressed growth and invasion of cancer cells is associated with up-regulation of let-7 microRNA.
Over-expression of cofilin-1 suppressed growth and invasion of cancer cells is associated with up-regulation of let-7 microRNA.
Biochimica et biophysica acta (2015-01-20)
Cheng-Han Tsai, Liang-Ting Lin, Chung-Yih Wang, Yu-Wen Chiu, Yen-Ting Chou, Shu-Jun Chiu, Hsin-Ell Wang, Ren-Shyan Liu, Chun-Yi Wu, Pei-Chia Chan, Muh-Hwa Yang, Shih-Hwa Chiou, Man-Jyun Liao, Yi-Jang Lee
ABSTRACT
Cofilin-1, a non-muscle isoform of actin regulatory protein that belongs to the actin-depolymerizing factor (ADF)/cofilin family is known to affect cancer development. Previously, we found that over-expression of cofilin-1 suppressed the growth and invasion of human non-small cell lung cancer (NSCLC) cells in vitro. In this study, we further investigated whether over-expression of cofilin-1 can suppress tumor growth in vivo, and performed a microRNA array analysis to better understand whether specific microRNA would be involved in this event. The results showed that over-expression of cofilin-1 suppressed NSCLC tumor growth using the xenograft tumor model with the non-invasive reporter gene imaging modalities. Additionally, cell motility and invasion were significantly suppressed by over-expressed cofilin-1, and down-regulation of matrix metalloproteinase (MMPs) -1 and -3 was concomitantly detected. According to the microRNA array analysis, the let-7 family, particularly let-7b and let-7e, were apparently up-regulated among 248 microRNAs that were affected after over-expression of cofilin-1 up to 7 days. Knockdown of let-7b or let-7e using chemical locked nucleic acid (LNA) could recover the growth rate and the invasion of cofilin-1 over-expressing cells. Next, the expression of c-myc, LIN28 and Twist-1 proteins known to regulate let-7 were analyzed in cofilin-1 over-expressing cells, and Twist-1 was significantly suppressed under this condition. Up-regulation of let-7 microRNA by over-expressed cofilin-1 could be eliminated by co-transfected Twist-1 cDNA. Taken together, current data suggest that let-7 microRNA would be involved in over-expression of cofilin-1 mediated tumor suppression in vitro and in vivo.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Gly-Gly, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Gly-Gly, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
D-Luciferin, synthetic
Sigma-Aldrich
D-Luciferin, synthetic, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
Gly-Gly, BioPerformance Certified, suitable for cell culture, ≥99%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)