Merck
  • The role of sulfur- and phosphorus-mobilizing bacteria in biochar-induced growth promotion of Lolium perenne.
The role of sulfur- and phosphorus-mobilizing bacteria in biochar-induced growth promotion of Lolium perenne.
FEMS microbiology ecology (2014-06-27)
Aaron Fox, Witold Kwapinski, Bryan S Griffiths, Achim Schmalenberger
ABSTRACT
Plants rely on microorganisms to mobilize organically and inorganically bound sulfur (S) and phosphorus (P) in which the plant can then readily utilize. The aim of this study was to investigate the role of S- and P-mobilizing bacteria in plant growth promotion in biochar-amended soil, which has been rarely investigated so far. Pot experiments of Lolium perenne were established on S and P limited soil with 1% or 2% biochar (Miscanthus × giganteus) or without biochar (control) for a period of 126 days. Both biochar amendments resulted in significant plant growth promotion. Rhizobacteria capable of growing with (1) S from aromatic sulfonates, (2) P from phosphate esters, (3) P from phosphonates, and (4) P from tri-calcium phosphates as sole source of S or P, respectively, were significantly more abundant in the biochar treatments. 16S rRNA gene-based rhizobacteria community analysis revealed a significant biochar treatment effect. Abundance of nematodes feeding on bacteria was also significantly increased in the biochar treatments. Diversity analysis of rhizospheric asfA and phnJ genes revealed broad sequence diversities in bacterial sulfonate and phosphonate-mineralizing capabilities. These findings suggest that biochar amendment enhances microbially mediated nutrient mobilization of S and P resulting in improved plant growth.
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard