Merck
Aldehyde oxidase activity in fresh human skin.
Drug metabolism and disposition: the biological fate of chemicals (2014-09-25)
Nenad Manevski, Kamal Kumar Balavenkatraman, Barbara Bertschi, Piet Swart, Markus Walles, Gian Camenisch, Hilmar Schiller, Olivier Kretz, Barbara Ling, Reto Wettstein, Dirk J Schaefer, Francois Pognan, Armin Wolf, Karine Litherland
ABSTRACT
Human aldehyde oxidase (AO) is a molybdoflavoenzyme that commonly oxidizes azaheterocycles in therapeutic drugs. Although high metabolic clearance by AO resulted in several drug failures, existing in vitro-in vivo correlations are often poor and the extrahepatic role of AO practically unknown. This study investigated enzymatic activity of AO in fresh human skin, the largest organ of the body, frequently exposed to therapeutic drugs and xenobiotics. Fresh, full-thickness human skin was obtained from 13 individual donors and assayed with two specific AO substrates: carbazeran and zoniporide. Human skin explants from all donors metabolized carbazeran to 4-hydroxycarbazeran and zoniporide to 2-oxo-zoniporide. Average rates of carbazeran and zoniporide hydroxylations were 1.301 and 0.164 pmol⋅mg skin(-1)⋅h(-1), resulting in 13 and 2% substrate turnover, respectively, after 24 hours of incubation with 10 μM substrate. Hydroxylation activities for the two substrates were significantly correlated (r(2) = 0.769), with interindividual variability ranging from 3-fold (zoniporide) to 6-fold (carbazeran). Inclusion of hydralazine, an irreversible inhibitor of AO, resulted in concentration-dependent decrease of hydroxylation activities, exceeding 90% inhibition of carbazeran 4-hydroxylation at 100 μM inhibitor. Reaction rates were linear up to 4 hours and well described by Michaelis-Menten enzyme kinetics. Comparison of carbazeran and zoniporide hydroxylation with rates of triclosan glucuronidation and sulfation and p-toluidine N-acetylation showed that cutaneous AO activity is comparable to tested phase II metabolic reactions, indicating a significant role of AO in cutaneous drug metabolism. To our best knowledge, this is the first report of AO enzymatic activity in human skin.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
p-Toluidine, for spectrophotometric det. of Au, Tl(III), W, ≥99.0%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
p-Toluidine, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
p-Toluidine, 99.6%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Zoniporide hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Carbazeran, ≥95% (HPLC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
USP
Triclosan, United States Pharmacopeia (USP) Reference Standard