Merck
  • Tanshinone IIA inhibits human gastric carcinoma AGS cell growth by decreasing BiP, TCTP, Mcl‑1 and Bcl‑xL and increasing Bax and CHOP protein expression.
Tanshinone IIA inhibits human gastric carcinoma AGS cell growth by decreasing BiP, TCTP, Mcl‑1 and Bcl‑xL and increasing Bax and CHOP protein expression.
International journal of molecular medicine (2014-10-02)
Chin-Cheng Su
ABSTRACT
Tanshinone IIA (Tan-IIA) is extracted from Danshen (Salviae Miltiorrhizae Radix) and is a natural anti-cancer agent, which possesses antitumor activity in a variety of human cancer cells. Tan-IIA can induce apoptosis and inhibit the proliferation of gastric cancer through different molecular mechanisms. However, the efficacy and molecular mechanism of Tan-IIA in gastric cancer have not been well studied. In the present study, the cytotoxicity of Tan-IIA in human gastric cancer AGS cells by 3-(4,5-dimethylthiazol-2-y1)-2,5-diphenyltetrazolium bromide assay was examined. The protein expression levels of B-cell lymphoma-extra large (Bcl-xL), Bcl-2-associated X protein (Bax), myeloid cell leukemia 1 protein (Mcl-1), translationally-controlled tumor protein (TCTP), binding immunoglobulin protein (BiP), calnexin, protein kinase-like endoplasmic reticulum kinase, eIF2α, activating transcription factor 4 (ATF4), inositol-requiring enzyme 1α (IRE1α), ATF6, caspase-12, caspase-9, caspase-3, C/EBP-homologous protein (CHOP) and β-actin in AGS cells were measured by western blot analysis. The results showed that Tan-IIA inhibited AGS cells in a time-and dose-dependent manner. AGS cells treated with Tan-IIA upregulated the protein expression of caspase-12, caspase-9, caspase--3, CHOP and Bax, but downregulated the protein expression of BiP, TCTP, Mcl-1 and Bcl-xL. These findings indicated that Tan-IIA inhibits the growth of human gastric cancer AGS cells. One of the molecular mechanisms may be through decreasing the protein expression of BiP to induce the activation of endoplasmic reticulum stress, followed by increasing the protein expression of caspase-12 to upregulate CHOP expression. The other may be through decreasing the protein expression of Mcl-1, Bcl-xL and TCTP, but increasing Bax, caspase-9 and caspase-3 to induce apoptosis. The chemotherapeutic potential of Tan-IIA for human gastric cancer warrants further study in the future.
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Supelco
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%