Merck
  • Design, synthesis and biological evaluation of nitro oxide donating N-hydroxycinnamamide derivatives as histone deacetylase inhibitors.
Design, synthesis and biological evaluation of nitro oxide donating N-hydroxycinnamamide derivatives as histone deacetylase inhibitors.
Chemical & pharmaceutical bulletin (2014-12-03)
Shiliang Tu, Hang Yuan, Jie Hu, Chengguang Zhao, Rui Chai, Hongfeng Cao
ABSTRACT
Novel nitro oxide (NO)-donating N-hydroxycinnamamide derivatives 12a-j were designed and synthesized by coupling the carboxyl group of N-hydroxycinnamamides with phenylsulfonylfuroxan through various diols or alkylol amines, and their in vitro biological activities were evaluated. It was discovered that most of target compounds showed good histone deacetylases (HDACs) inhibition and anti-tumor activities, particularly for 12j, which had great HDACs inhibitory activities (IC50s=0.15-0.26 µM) and antiproliferative effects (IC50s=3.21-7.12 µM) comparable to suberoylanilide hydroxamic acid (SAHA) (IC50s=0.16-1.41 µM for HDACs, IC50s=3.15-7.45 µM for cancer cell inhibition). Furthermore, compound 12j with strong antitumor activities produced high levels of NO (up to 8.0 µM of nitrites/nitrates) in colon cancer cells, and its antiproliferative activity was nearly half-diminished by hemoglobin (10 µM), an NO scavenger. These results suggest that the strong antiproliferative activity of 12j could be attributed to the additive effects of high levels of NO production and inhibition of HDAC in the cancer cells.
Product Description
Supelco
Tetramethylsilane, analytical standard, suitable for NMR (spectroscopy), ACS reagent
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Tetramethylsilane, NMR grade, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard