Merck
  • Effects of raspberry fruit extracts and ellagic acid on respiratory burst in murine macrophages.
Effects of raspberry fruit extracts and ellagic acid on respiratory burst in murine macrophages.
Food & function (2014-04-05)
Lina Raudone, Ramune Bobinaite, Valdimaras Janulis, Pranas Viskelis, Sonata Trumbeckaite
ABSTRACT
The mechanism of action of polyphenolic compounds is attributed to their antioxidant, anti-inflammatory, and anti-proliferative properties and their effects on subcellular signal transduction, cell cycle impairment and apoptosis. A raspberry (Rubus idaeus L.) fruit extract contains various antioxidant active compounds, particularly ellagic acid (EA); however the exact intracellular mechanism of their action is not fully understood. The aim of the study was to evaluate the antioxidant effect of raspberry extracts, and that of ellagic acid by assessment of the production of the reactive oxygen species (ROS) by murine macrophage J774 cells. Raspberry extracts and their active compound EA did not affect or had very minor effects on cell viability. No significant difference in the ROS generation in arachidonic acid stimulated macrophages was determined for raspberry extracts and EA whereas in the phorbol-12 myristate-13 acetate model ROS generation was significantly (p < 0.05) reduced. Our observation that raspberry pomace extracts in vitro reduce ROS production in a J774 macrophage culture suggests that raspberry extract and ellagic acid mediated antioxidant effects may be due to the regulation of NADPH oxidase activity.
Product Description
Supelco
Ellagic acid, analytical standard
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium carbonate, powder, 99.999% trace metals basis
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC