Merck
  • Toll-like receptor 4 regulates platelet function and contributes to coagulation abnormality and organ injury in hemorrhagic shock and resuscitation.
Toll-like receptor 4 regulates platelet function and contributes to coagulation abnormality and organ injury in hemorrhagic shock and resuscitation.
Circulation. Cardiovascular genetics (2014-07-23)
Ning Ding, Guoqiang Chen, Rosemary Hoffman, Patricia A Loughran, Chhinder P Sodhi, David J Hackam, Timothy R Billiar, Matthew D Neal
ABSTRACT
Growing evidence indicates that the presence of toll-like receptor 4 (TLR4) on platelets is a key regulator of platelet number and function. Platelets exposed to TLR4 agonists may serve to activate other cells such as neutrophils and endothelial cells in sepsis and other inflammatory conditions. The functional significance of platelet TLR4 in hemorrhagic shock (HS), however, remains unexplored. Using thromboelastography and platelet aggregometry, we demonstrate that platelet function is impaired in a mouse model of HS with resuscitation. Further analysis using cellular-specific TLR4 deletion in mice revealed that platelet TLR4 is essential for platelet activation and function in HS with resuscitation and that platelet TLR4 regulates the development of coagulopathy after hemorrhage and resuscitation. Transfusion of TLR4-negative platelets into mice resulted in protection from coagulopathy and restored platelet function. Additionally, platelet-specific TLR4 knockout mice were protected from lung and liver injury and exhibited a marked reduction in systemic inflammation as measured by circulating interleukin-6 after HS with resuscitation. We demonstrate for the first time that platelet TLR4 is an essential mediator of the inflammatory response as well as platelet activation and function in HS and resuscitation.
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Prostaglandin E1, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Alprostadil, European Pharmacopoeia (EP) Reference Standard
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant