Merck
  • Microvascular lesions by estrogen-induced ID3: its implications in cerebral and cardiorenal vascular disease.
Microvascular lesions by estrogen-induced ID3: its implications in cerebral and cardiorenal vascular disease.
Journal of molecular neuroscience : MN (2014-08-19)
Jayanta K Das, Quentin Felty
ABSTRACT
Severe symptoms of cerebral and cardiorenal vascular diseases can be triggered when cerebral, coronary, or glomerular arterioles grow inappropriately as a result of abnormal cell proliferation. The risk factor(s) and molecular mechanisms responsible for microvascular lesion formation are largely unknown. Although controversial, both animal and epidemiological studies have shown that estrogen increases the risk of stroke which may be due to microvascular lesions. Since microvascular diseases are characterized by excessive vessel growth, it is plausible that estrogen-induced neovascularization contributes to the growth of microvascular lesions. We present evidence for how ID3 overexpression in endothelial cells contributes to the development of an estrogen-induced neovascular phenotype with an additional focus on Pyk2 kinase. Our data showed that ID3 overexpression increased neovascularization, cell migration, and spheroid growth of human cerebral microvascular endothelial cells, hCMEC/D3. ID3-overexpressing cells showed significant estrogen-induced G2/M phase transition. Estrogen treatment increased both ID3 phosphorylation; total protein that was inhibited by tamoxifen, and Pyk2-mediated estrogen-induced ID3 mRNA expression. These findings suggest that Pyk2 signals ID3 expression and ID3 is necessary for estrogen-induced neovascularization in hCMEC/D3 cells. A better understanding of how microvascular lesions depend on ID3 may open new avenues for prevention and treatment of neurological diseases.
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%