Merck
  • Mucoadhesive polymers-based film as a carrier system for sublingual delivery of glutathione.
Mucoadhesive polymers-based film as a carrier system for sublingual delivery of glutathione.
The Journal of pharmacy and pharmacology (2014-10-11)
Guanyu Chen, Craig Bunt, Jingyuan Wen
ABSTRACT
The sublingual mucosa provides a promising route for the delivery of glutathione (GSH). However, challenges are encountered in developing sublingual mucoadhesive drug delivery formulations such as: prolonging drug retention, uniform drug content, desirable drug release profiles, adequate drug permeation and efficient delivery of GSH. The aim of this study was to develop a suitable mucoadhesive polymer-based sublingual film. The mucoadhesive films were prepared by casting method. Several characterization studies including thickness, weight uniformity, surface pH, elongation, mucoadhesiveness, swelling and erosion were carried out on preliminary formulations to optimise formulations for in-vitro drug release and ex-vivo permeation studies. The optimal mucoadhesive polymer-based films showed acceptable physical properties and good mucoadhesion, and remained attached to excised porcine sublingual mucosa for sufficient time, providing a sustained delivery of GSH through the mucosal epithelial. The optimal mucoadhesive films may provide a promising drug delivery platform to develop commercial sublingual products of GSH as well as a wide range of protein and peptide drugs.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%