Merck
  • Simultaneous determination of polar pharmaceuticals and personal care products in biological organs and tissues.
Simultaneous determination of polar pharmaceuticals and personal care products in biological organs and tissues.
Journal of chromatography. A (2014-06-25)
Rumi Tanoue, Kei Nomiyama, Haruna Nakamura, Terutake Hayashi, Joon-Woo Kim, Tomohiko Isobe, Ryota Shinohara, Shinsuke Tanabe
ABSTRACT
In the present study, a sensitive and accurate isotope dilution method was developed for the simultaneous determination of 17 polar pharmaceutical and personal care product (PPCP) residues (logKow=1.40-5.74), including 14 pharmaceuticals and 3 personal care products, in biological organs and tissues. The proposed method involved enzymatic hydrolysis, followed by sequential clean-up using silica gel chromatography and gel permeation chromatography, and analysis via ultra-high-performance liquid chromatography with tandem mass spectrometry. This method yielded acceptable absolute recoveries (48-88%) and internal standard-corrected recoveries (90-130%) for 17 PPCPs. Method detection limits were between 0.0092 and 3.2ngg(-1) wet weight, and the limits of quantification were between 0.020 and 8.7ngg(-1) wet weight. The method can be used to readily detect the target compounds at trace levels while minimizing the required sample volume. The developed method was applied to the determination of 17 PPCPs in the liver and kidney of 17 birds collected from Japan and also in the plasma, liver, and brain of 7 cyprinoid fish from an effluent-dominated stream in Japan. Triclosan was detected in 5 of 11 fish-eating birds but not in non-fish-eating birds, suggesting the contamination of prey fish by the chemical. Non-steroidal anti-inflammatory drugs, antibacterial agents, and psychotropic agents were frequently detected in the fish tissues. In addition, 7 of the target compounds were found in fish brain. The median brain/plasma ratios of the psychotropic agents ranged from 1.6 (carbamazepine) to 12 (diphenhydramine), indicating high transportability to fish brain.
Product Description
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Ethoxybenzamide, 97%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Supelco
Acetonitrile, analytical standard
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer