Merck
  • Simple analytical strategy for MALDI-TOF-MS and nanoUPLC-MS/MS: quantitating curcumin in food condiments and dietary supplements and screening of acrylamide-induced ROS protein indicators reduced by curcumin.
Simple analytical strategy for MALDI-TOF-MS and nanoUPLC-MS/MS: quantitating curcumin in food condiments and dietary supplements and screening of acrylamide-induced ROS protein indicators reduced by curcumin.
Food chemistry (2014-12-23)
Yu-Shu Huang, Tusty-Jiuan Hsieh, Chi-Yu Lu
ABSTRACT
Curcumin is the major active ingredient of turmeric and is widely used as a preservative, flavouring and colouring agent. Curcumin is a potent substance with several functions, including antioxidant, antitumor, anti-inflammatory, antimicrobial, antiparasitic, antimutagenic, chemopreventive and chemotherapeutic activities. Matrix-assisted laser desorption/ionisation coupled with time-of-flight mass spectrometry (MALDI-TOF-MS) has been used to analyse various molecules (including natural antioxidants). This study established an expeditious method that couples MALDI-TOF-MS with a simple dilution method to quantify curcumin in food condiments and dietary supplements. The linear range of curcumin detection ranged from 1 to 100 μg/mL. In further experiments, liver cells were treated with curcumin after exposure to acrylamide. Nano ultra performance liquid chromatographic system (nanoUPLC) coupled with tandem mass spectrometry (MS/MS) was used to evaluate the potential proteins and protein modifications induced by acrylamide. The results indicate that curcumin reduces the effects of reactive oxygen species induced by acrylamide.
Product Description
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Supelco
Ofloxacin, VETRANAL®, analytical standard
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
Thiosalicylic acid, 97%
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, Ultra pure
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard