Merck
  • Honeybee venom possesses anticancer and antiviral effects by differential inhibition of HPV E6 and E7 expression on cervical cancer cell line.
Honeybee venom possesses anticancer and antiviral effects by differential inhibition of HPV E6 and E7 expression on cervical cancer cell line.
Oncology reports (2015-01-31)
Yong-Wan Kim, Pankaj Kumar Chaturvedi, Sung Nam Chun, Yang Gu Lee, Woong Shick Ahn
ABSTRACT
Bee venom (BV) therapy is a type of alternative medical treatment used to treat various diseases in oriental medicine. The mechanisms underlying the effects of BV remain poorly understood. In the present study, we evaluated the antiviral effect of BV on cervical carcinoma cell lines (CaSki, HeLa, C33A and TC-1). BV treatments resulted in a more significant suppression of cell growth in HPV 16-infected cells (CaSki) and a lesser suppression in HPV 18-infected cells (HeLa). However, less suppression was observed in HPV-negative C33A cells. In 10 µg/ml BV-treated CaSki cells, the mRNA expression and protein levels of HPV16 E6 and E7 were significantly decreased by BV, while HPV18 E6 and E7 mRNA expression levels were not significantly altered by 10 µg/ml BV-treated HeLa cells. The antitumor effects of BV were in accordance with in vitro data, in restricting tumor growth in vivo and were much more effective on the suppression of tumor growth. Furthermore, the mRNA and protein expression levels of HPV16 E6 and E7 were decreased by BV in TC-1 tumors. These findings demonstrated the antiviral effects of BV in HPV-infected cervical cancer cells and the anticancer effects of BV in HPV16 E6/E7-expressed TC-1 tumors. Collectively, BV plays a differential role in suppressing HPV16-infected cells (CaSki cells) and HPV18-infected cells (HeLa cells) by the downregulation of E6/E7 protein of HPV16/18.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)