Merck
  • Characterization of the neuroprotective potential of derivatives of the iron chelating drug deferiprone.
Characterization of the neuroprotective potential of derivatives of the iron chelating drug deferiprone.
Neurochemical research (2015-01-07)
Pamela Maher, George J Kontoghiorghes
ABSTRACT
There is growing evidence for alterations in iron homeostasis during aging that are exacerbated in neurodegenerative diseases such as Alzheimer's disease. However, since essentially all neurodegenerative diseases are multi-factorial in the sense that there are a large number of mechanisms that can be identified as contributing to nerve cell death, iron chelators that have additional activities might be the most useful for the treatment of age-related CNS diseases. We have described a series of cell culture-based assays that define molecular toxicity pathways relevant to neurodegenerative diseases and have used these assays to identify potential therapeutic compounds for the treatment of these diseases. Deferiprone is a blood brain barrier permeable, low molecular weight iron chelator that has been used for many years to treat systemic iron disease. In this study, we describe the use of our cell culture-based screening assays to identify deferiprone derivatives with the greatest therapeutic potential for the treatment of CNS diseases. We show that several derivatives are much more potent than deferiprone at reducing oxidative stress and preventing nerve cell death induced by multiple, age-related insults. In addition, we show that both deferiprone and the derivatives modulate several distinct signaling pathways associated with neuroprotection. All of the compounds were able to both inhibit the activation of p38 MAP kinase and JNK kinase and prevent the loss of PI3 kinase activity in response to a toxic stress. These results strongly suggest that these compounds have significant potential for the treatment of CNS diseases.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, ≥98.0%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
SAFC
BIS-TRIS
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O