Merck
  • Gastric vagal motoneuron function is maintained following experimental spinal cord injury.
Gastric vagal motoneuron function is maintained following experimental spinal cord injury.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2014-10-16)
E M Swartz, G M Holmes
ABSTRACT
Clinical reports indicate that spinal cord injury (SCI) initiates profound gastric dysfunction. Gastric reflexes involve stimulation of sensory vagal fibers, which engage brainstem circuits that modulate efferent output back to the stomach, thereby completing the vago-vagal reflex. Our recent studies in a rodent model of experimental high thoracic (T3-) SCI suggest that reduced vagal afferent sensitivity to gastrointestinal (GI) stimuli may be responsible for diminished gastric function. Nevertheless, derangements in efferent signals from the dorsal motor nucleus of the vagus (DMV) to the stomach may also account for reduced motility. We assessed the anatomical, neurophysiological, and functional integrity of gastric-projecting DMV neurons in T3-SCI rats using: (i) retrograde labeling of gastric-projecting DMV neurons; (ii) whole cell recordings from gastric-projecting neurons of the DMV; and, (iii) in vivo measurements of gastric contractions following unilateral microinjection of thyrotropin-releasing hormone (TRH) into the DMV. Immunohistochemical analysis of gastric-projecting DMV neurons demonstrated no difference between control and T3-SCI rats. Whole cell in vitro recordings showed no alteration in DMV membrane properties and the neuronal morphology of these same, neurobiotin-labeled, DMV neurons were unchanged after T3-SCI with regard to cell size and dendritic arborization. Central microinjection of TRH induced a significant facilitation of gastric contractions in both control and T3-SCI rats and there were no significant dose-dependent differences between groups. Our data suggest that the acute, 3 day to 1 week post-SCI, dysfunction of vagally mediated gastric reflexes do not include derangements in the efferent DMV motoneurons.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Nickel(II) sulfate, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Cobalt(II) chloride, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Cobalt(II) chloride, 97%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material