Merck
  • Blockade of spinal glutamate recycling produces paradoxical antinociception in rats with orofacial inflammatory pain.
Blockade of spinal glutamate recycling produces paradoxical antinociception in rats with orofacial inflammatory pain.
Progress in neuro-psychopharmacology & biological psychiatry (2014-12-03)
Kui Y Yang, Jun H Mun, Ki D Park, Min J Kim, Jin S Ju, Seong T Kim, Yong C Bae, Dong K Ahn
ABSTRACT
In our current study, we investigated the role of spinal glutamate recycling in the development of orofacial inflammatory pain. DL-threo-β-benzyloxyaspartate (TBOA) or methionine sulfoximine (MSO) was administered intracisternally to block spinal glutamate transporter and glutamine synthetase activity in astroglia. Intracisternal administration of high dose TBOA (10 μg) produced thermal hyperalgesia in naïve rats but significantly attenuated the thermal hyperalgesia in rats that had been pretreated with interleukin (IL)-1β or Complete Freund's Adjuvant (CFA). In contrast, intracisternal injection of MSO produced anti-hyperalgesic effects against thermal stimuli in CFA-treated rats only. To confirm the paradoxical antinociceptive effects of TBOA and MSO, we examined changes in c-Fos expression in the medullary dorsal horn produced by thermal stimulation in naïve, IL-1β-, or CFA-treated rats, after intracisternal injections of TBOA and MSO. Intracisternal administration of TBOA significantly increased c-Fos immunoreactivity in naïve rats. In contrast, intracisternal administration of TBOA significantly decreased the up-regulation of c-Fos immunoreactivity in the medullary dorsal horn of IL-1β- and CFA-treated rats. However, intracisternal injection of MSO blocked the up-regulation of c-Fos immunoreactivity in CFA-treated rats only. We also investigated the effects of botulinum toxin type A (BoNT-A) on TBOA-induced paradoxical antinociception in CFA-treated rats, as BoNT-A inhibits the release of neurotransmitters, including glutamate. BoNT-A treatment reversed behavioral responses produced by intracisternal administration of TBOA in CFA-treated rats. These results suggest that the paradoxical responses produced by blocking glutamate transporters under inflammatory pain conditions are mediated by the modulation of glutamate release from presynaptic terminals. Moreover, blockade of glutamate reuptake could represent a new therapeutic target for the treatment of chronic inflammatory pain conditions.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized