Merck
  • Safflower yellow inhibits angiotensin II-induced adventitial fibroblast proliferation and migration.
Safflower yellow inhibits angiotensin II-induced adventitial fibroblast proliferation and migration.
Journal of pharmacological sciences (2014-09-19)
Yueqiang Liu, Xuefeng Tian, Mingzhu Cui, Suzhen Zhao
ABSTRACT
Safflower yellow (SY) has been widely used in Chinese medicine for the treatment of ischemic cardiocerebrovascular disease. Recent studies have indicated that SY has a reverse effect on vascular remodeling (VR). However, its detailed mechanisms require further study to provide more scientific evidence for the clinical treatment of VR. This study aims to investigate the effects of SY on angiotensin II (Ang II)-induced cell proliferation, migration, apoptosis, and extracellular matrix in rat aortic adventitial fibroblasts (AFs). The proliferation and migration rates of AFs treated with Ang II for 24 h were higher than those of untreated AFs; and increases in the expression of p-ERK1/2, AP-1, collagen I, and collagen III were observed. Treatment with SY significantly downregulated cell proliferation, migration, and the expression of p-ERK1/2, AP-1, collagen I, and collagen III. We also found that the cell percentage of apoptosis of AFs treated with Ang II for 24 h was lower than those of untreated AFs. After treatment with SY, the percentage of apoptosis was increased. SY exhibits anti-proliferative, anti-migratory, and pro-apoptotic activities in rat aortic AFs, perhaps through the Ang II/ERK/AP-1 signaling pathway. The present findings may provide new clues regarding the potential function of SY to treat or prevent VR.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%