Merck
  • Peroxiredoxin 2 is involved in vasculogenic mimicry formation by targeting VEGFR2 activation in colorectal cancer.
Peroxiredoxin 2 is involved in vasculogenic mimicry formation by targeting VEGFR2 activation in colorectal cancer.
Medical oncology (Northwood, London, England) (2014-12-05)
Shouru Zhang, Zhongxue Fu, Jinlai Wei, Jinbao Guo, Maoxi Liu, Kunli Du
ABSTRACT
The mammalian peroxiredoxin 2 (Prdx2) is a member of thiol-dependent antioxidant proteins and plays an important role in the progression of colorectal cancer (CRC). The aim of this study was to confirm the role of Prdx2 in formation of VM and progression of CRC. Immunohistochemistry and CD34/periodic acid Schiff double staining were performed to explore the expression of Prdx2 and VM formation in 70 CRC tissues, and there was a positive correlation between Prdx2 expression and VM formation by the Pearson correlation coefficient (r = 0.282, p < 0.05). Prdx2 was suppressed in poorly differentiated HCT116 cells by Prdx2-siRNA-LV transduction. The expression of Prdx2 at both mRNA and protein levels in HCT116 cells transfected with the Prdx2 siRNA was significantly lower than that of negative control siRNA as confirmed by quantitative real-time PCR and Western blotting analysis, respectively (p < 0.05). The well-established in vitro 3D culture model was chosen to investigate the VM formation of HCT116 cells. The numbers of the tubular structures were significantly fewer in Prdx2 siRNA explants than those of negative control siRNA explants after VEGF induction (p < 0.05). Although VEGFR2 expressions had no change after VEGF induction, we found that VEGFR2 phosphorylation levels were markedly reduced in cells of siPrdx2 over time compared with those of negative control siRNA by Western blotting analysis (p < 0.05, p < 0.01). The effects of Prdx2 siRNA on the invasive capabilities of HCT116 cells with VEGF induction were examined by using Matrigel invasion chamber assay. The invasive capabilities of HCT116 cells were significantly declined in Prdx2 siRNA explants than those of negative control siRNA explants (p < 0.05). The effects of Prdx2 siRNA on pathological tumor growth were examined by using a tumor xenograft model in vivo. After implant of HCT116 cells that transduced with Prdx2 siRNA and negative control siRNA as xenografts into nude mice, the growth of xenograft tumors with Prdx2 siRNA was much slower than that of negative control siRNA, and the volumes of tumor xenografts with Prdx2 siRNA were smaller than those of negative control siRNA after 5 weeks (p < 0.05). Further conclusion showed that Prdx2 regulates VM formation by targeting VEGFR2 activation, which now represents as a therapeutic target for RC.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
1,5,7-Triazabicyclo[4.4.0]dec-5-ene, 98%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.