Merck
  • Atorvastatin administered before myocardial infarction in rats improves contractility irrespective of metabolic changes.
Atorvastatin administered before myocardial infarction in rats improves contractility irrespective of metabolic changes.
Clinical and experimental pharmacology & physiology (2014-09-17)
Tatiana Ederich Lehnen, Alexandre Machado Lehnen, Angela Maria Vicente Tavares, Adriane Belló-Klein, Melissa Medeiros Markoski, Ubiratan Fabres Machado, Beatriz Schaan
ABSTRACT
Statins have a beneficial effect after myocardial infarction, but the relationship between glucose transporters and their use before the event has not yet been studied. We assessed the effects of atorvastatin treatment pre- and post-myocardial infarction on cardiovascular function and glucose transporter 4 (GLUT4) in the heart. Wistar-Kyoto rats were treated with 20 mg/kg atorvastatin or vehicle for 14 days before coronary artery occlusion surgery (myocardial infarction) or sham surgery. Echocardiographic evaluations were carried out 48 h after myocardial infarction (protocol A) and after 7 days (protocol B), when atorvastatin was also administered. Plasma inflammatory markers and GLUT4 in the heart were also evaluated. Animals were divided into the following groups: sham-operated and vehicle (C), myocardial infarction and vehicle (I), sham-operated and atorvastatin (CAt) and myocardial infarction and atorvastatin (IAt). After 48 h, myocardial infarction induced higher left ventricular fractional shortening in IAt versus I (~ 60%, P = 0.036), and the ejection fraction was lower (protocol A ~ 37%; protocol B ~ 30%). Myocardial infarction was associated with a rise in plasma membrane GLUT4 after 48 h (~ 40%, P < 0.001), and a reduction in GLUT4 after 7 days (I 25%; IAt 49%, P < 0.001). Atorvastatin treatment for 48 h after the infarction did not change GLUT4 expression, and after 7 days it had an additional negative effect on GLUT4 content (~ 39%, P = 0.030). In conclusion, atorvastatin treatment pre- and post-myocardial infarction improved myocardial contractility after 48 h, but not after 7 days, and was not associated with an increase in GLUT4 expression.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri