Merck
  • Hyperoxia resensitizes chemoresistant glioblastoma cells to temozolomide through unfolded protein response.
Hyperoxia resensitizes chemoresistant glioblastoma cells to temozolomide through unfolded protein response.
Anticancer research (2014-06-13)
Derek Lee, Stella Sun, Amy S W Ho, Karrie M Y Kiang, Xiao Qin Zhang, Fei Fan Xu, Gilberto K K Leung
ABSTRACT
Intratumoural hypoxia is associated with chemoresistance in glioblastoma multiforme (GBM), a highly malignant brain tumour. Adaptive response to endoplasmic reticulum stress induced by temozolomide is a major obstacle in recurrent GBM. We investigated whether hyperoxia resensitizes temozolomide-resistant GBM cells to temozolomide by abrogating the hypoxia-induced, unfolded protein response (UPR)-related protective mechanisms. We examined changes to key UPR modulators in temozolomide-sensitive and -resistant human GBM cells (D54 and U87) treated with/without temozolomide at different oxygen concentrations using western blotting, and cytotoxic benefits of overexpressing key chaperone, P4HB, in GBM cells (U87 and U251) under normoxia and hyperoxia. Hyperoxia, alone or synergistically with temozolomide, activated the UPR in sensitive and resistant D54 and U87 cell lines. Hyperoxia also reduced survival benefit of U87 and U251 cells with P4HB overexpression through the UPR. Hyperoxia enhanced GBM cell sensitivity to temozolomide, likely through UPR, highlighting an important treatment modality targeting chemosensitive and -resistant GBM.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material