Merck
  • Anti-inflammatory effects of methyl ursolate obtained from a chemically derived crude extract of apple peels: potential use in rheumatoid arthritis.
Anti-inflammatory effects of methyl ursolate obtained from a chemically derived crude extract of apple peels: potential use in rheumatoid arthritis.
Archives of pharmacal research (2014-04-16)
Tatiana A Pádua, Bianca S S C de Abreu, Thadeu E M M Costa, Marcos J Nakamura, Lígia M M Valente, Maria das Graças Henriques, Antonio C Siani, Elaine C Rosas
ABSTRACT
Ursolic acid (UA), a pentacyclic triterpene acid found in apple peels (Malus domestica, Borkh, Rosaceae), has a large spectrum of pharmacological effects. However, the vegetal matrix usually produces highly viscous and poorly soluble extracts that hamper the isolation of this compound. To overcome this problem, the crude EtOH-AcOEt extract of commercial apple peels was exhaustively treated with diazomethane, after which methyl ursolate (MU) was purified by column chromatography and characterized spectrometrically. The anti-inflammatory effects of UA and MU (50 mg/kg) were analyzed by zymosan-induced paw edema, pleurisy and in an experimental arthritis model. After 4 h of treatment with UA and MU, paw edema was reduced by 46 and 44 %, respectively. Both UA and MU inhibited protein extravasation into the thoracic cavity; tibio-femoral edema by 40 and 48 %, respectively; and leukocyte influx into the synovial cavity after 6 h by 52 and 73 %, respectively. Additionally, both UA and MU decreased the levels of mediators related to synovial inflammation, such as KC/CXCL-1 levels by 95 and 90 %, TNF-α levels by 76 and 71 %, and IL-1β levels by 57 and 53 %, respectively. Both the compounds were equally effective when assayed in different inflammatory models, including experimental arthritis. Hence, MU may be considered to be a useful anti-inflammatory derivative to overcome the inherent poor solubility of UA for formulating pharmaceutical products.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology