Merck
  • High-level expression, purification and characterization of active human C1q and tumour necrosis factor-related protein-1 in Escherichia coli.
High-level expression, purification and characterization of active human C1q and tumour necrosis factor-related protein-1 in Escherichia coli.
Letters in applied microbiology (2014-05-13)
H Li, X Hui, K Li, X Tang, X Hu, A Xu, D Wu
ABSTRACT
C1q and tumour necrosis factor-related proteins (CTRPs) are a family of adiponectin paralogues. CTRP1 plays important biological functions in diabetes, obesity and hypertension. To further explore the physiological roles of human CTRP1 and its mechanisms of action, hCTRP1 gene was expressed in Escherichia coli. In the E. coli expression system, a large amount of soluble thioredoxin (Trx)-hCTRP1 fusion protein could be produced using the expression plasmid pET32a (+) and induction with IPTG at 18°C, which accounts about 20% of the total soluble bacterial proteins. The recombinant Trx-hCTRP1 fusion protein was purified to an approx. 95% purity using Ni-NTA affinity chromatography and Superdex G-75 column with a yield of about 28-mg protein from 1-l bacterial cultures. The purified recombinant Trx-hCTRP1 was shown to be active under in vivo and in vitro assay conditions. CTRP1 plays important biological functions and warrants further investigation. However, large-scale production of recombinant CTRP1 has been technically challenging, which becomes a major obstacle in the structural and functional analysis of this important family of proteins. To explore the possible clinical applications and mechanisms of its action, an efficient method to produce large amounts of active recombinant human CTRP1 is necessary. This study should facilitate basic functional and pharmacological studies of this important protein family.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%