Merck
Interactions of Streptococcus iniae with phagocytic cell line.
Microbes and infection (2014-06-24)
Fatima El Aamri, S Remuzgo-Martínez, Félix Acosta, Fernando Real, José Ramos-Vivas, José M Icardo, Daniel Padilla
ABSTRACT
Streptococcus iniae has become one of the most serious aquatic pathogens in the last decade, causing large losses in wild and farmed fish worldwide. There is clear evidence that this pathogen is capable not only of causing serious disease in fish but also of being transferred to and infecting humans. In this study, we investigate the interaction of S. iniae with two murine macrophage cell lines, J774-A1 and RAW 264.7. Cytotoxicity assay demonstrated significant differences between live and UV-light killed IUSA-1 strains. The burst respiratory activity decreased to baseline after 1 and 4 h of exposure for J774-A1 and RAW 264.7, respectively. Immunofluorescent and ultrastructural study of infected cells confirmed the intracellular localization of bacteria at 1 h and 24 h post-infection. Using qRT-PCR arrays, we investigated the changes in the gene expression of immune relevant genes associated with macrophage activation. In this screening, we identified 11 of 84 genes up-regulated, we observed over-expression of pro-inflammatory response as IL-1α, IL-1β, and TNF-α, without a good anti-inflammatory response. Present findings suggest a capacity of S. iniae to modulate a mammalian macrophages cell lines to their survival and replication intracellular, which makes this cell type as a reservoir for continued infection.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)