Merck
  • Antigenotoxic and antioxidant effects of the Mongolian medicinal plant Leptopyrum fumarioides (L): an in vitro study.
Antigenotoxic and antioxidant effects of the Mongolian medicinal plant Leptopyrum fumarioides (L): an in vitro study.
Journal of ethnopharmacology (2014-06-15)
Delgerbat Boldbaatar, Hesham R El-Seedi, Meriana Findakly, Safa Jabri, Batkhuu Javzan, Battsetseg Choidash, Ulf Göransson, Björn Hellman
ABSTRACT
Leptopyrum fumarioides has been used in the traditional medicine of Mongolia for the treatment of various diseases, including drug intoxications. However, since there is only sparse information about its chemistry, active components, and pharmacological and toxicological effects, the major aim of the present study employing mouse lymphoma cells was to evaluate the genotoxic and antigenotoxic/antioxidative effects of extracts and components isolated from this plant. A crude methanol extract was separated into three different sub-extracts: dichloromethane, n-butanol, and water. The major constituent of the n-butanol extract, i.e., the flavone luteolin-7-O-glucoside and a mixture of the most abundant compounds in the dichloromethane sub-extract were then isolated. DNA damage was evaluated using the comet assay; the antioxidant activity was evaluated using the DPPH radical scavenging assay. The crude methanol extract, the dichloromethane sub-extract and the mixture of compounds isolated from the latter fraction, increased the level of DNA damage after three hours of exposure. In contrast, no increase in DNA damage was observed in the cells that had been exposed to the n-butanol and water sub-extracts, or to the pure flavone. When non-DNA damaging concentrations of extracts and compounds were tested together with the DNA damaging agent catechol, all sub-extracts were found to reduce the catechol-induced DNA damage (the flavone was then found to be the most effective protective agent). The n-butanol sub-extract and the flavone were also found to have the most prominent antioxidative effects. Based on the results from the present study, components in Leptopyrum fumarioides were found to protect the DNA damage induced by catechol, probably by acting as potent antioxidants.
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrocatechol, ≥99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture