Merck
  • Plumbagin suppresses tumor cell growth in oral squamous cell carcinoma cell lines.
Plumbagin suppresses tumor cell growth in oral squamous cell carcinoma cell lines.
Oral diseases (2015-01-13)
T Ono, A Ota, K Ito, T Nakaoka, S Karnan, H Konishi, A Furuhashi, T Hayashi, Y Yamada, Y Hosokawa, Y Kazaoka
ABSTRACT
Plumbagin (PL), a naturally occurring quinoid, exerts antitumoral effects in diverse types of cancer cells. However, the effect of PL on tumor cell proliferation in oral squamous cell carcinoma (OSCC) remains poorly understood. In this study, we assessed the efficacy of PL, in human OSCC cells. The effect of PL on the cell growth and apoptosis of OSCC cell lines was evaluated using MTT and Annexin V assays, respectively. The effect of PL on mitochondrial membrane potential loss and reactive oxygen species (ROS) generation was evaluated using flow cytometry analysis. MTT assay showed that PL dose-dependently suppressed OSCC cell growth, with IC50 values ranging from 3.87 to 14.6 μM. Flow cytometry analysis revealed that PL treatment resulted in a significant decrease in mitochondrial membrane potential and an increase in the number of apoptotic cells. Notably, ROS generation was significantly elevated after PL treatment. Furthermore, a ROS scavenger, N-acetylcysteine (NAC), clearly suppressed the decrease in mitochondrial membrane potential, increase of caspase-3/7 activity, and apoptosis after PL treatment. This study provides the considerable evidence of the tumor-suppressive effect of PL, thereby highlighting its therapeutic potential for OSCC treatment.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%