Merck
  • RXR agonist modulates TR: corepressor dissociation upon 9-cis retinoic acid treatment.
RXR agonist modulates TR: corepressor dissociation upon 9-cis retinoic acid treatment.
Molecular endocrinology (Baltimore, Md.) (2014-12-30)
Juliana Fattori, Jéssica L O Campos, Tábata R Doratioto, Lucas M Assis, Mariela T Vitorino, Igor Polikarpov, José Xavier-Neto, Ana Carolina M Figueira
ABSTRACT
Transcriptional regulation controlled by thyroid hormone receptor (TR) drives events such as development, differentiation, and metabolism. TRs may act either as homodimers or as heterodimers with retinoid X receptor (RXR). Thyroid hormone T3 preferentially binds TR-RXR heterodimers, which activate transcription through coactivator recruitment. However, it is unclear whether TR-RXR heterodimers may also be responsive to the canonical RXR agonist 9-cis retinoic acid (9C) in the context of physiological gene regulation. New structural studies suggest that 9C promotes the displacement of bound coactivators from the heterodimer, modifying TR-RXR activity. To shed light on the molecular mechanisms that control TR-RXR function, we used biophysical approaches to characterize coregulator recruitment to TR-TR or to TR-RXR in the presence of T3 and/or 9C as well as cell-based assays to establish the functional significance of biophysical findings. Using cell-based and fluorescence assays with mutant and wild-type TR, we show that 9C does indeed have a function in the TR-RXR heterodimer context, in which it induces the release of corepressors. Furthermore, we show that 9C does not promote detectable conformational changes in the structure of the TR-RXR heterodimer and does not affect coactivator recruitment. Finally, our data support the view that DNA binding domain and Hinge regions are important to set up NR-coactivator binding interfaces. In summary, we showed that the RXR agonist 9C can regulate TR function through its modulation of corepressor dissociation.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)