Merck
  • Mechanism of chemical activation of sodium chloride in the presence of amino acids.
Mechanism of chemical activation of sodium chloride in the presence of amino acids.
Food chemistry (2014-07-24)
Anja K K Rahn, Varoujan A Yaylayan
ABSTRACT
Sodium chloride has been shown to promote chlorination of glycerol during thermal processing. However, the detailed mechanism of this reaction is not well understood. Preliminary experiments have indicated that the reaction mixture should contain an amino acid and it should be dissolved thoroughly in water in order to induce chlorination. These observations are consistent with the process of dissociation of sodium chloride and its re-association with amino acid and eventual formation of the chlorinating agent in the form of the hydrochloride salt. Release of HCl from this salt can be manifested in chlorination and hydrolytic reactions occurring during thermal processing. The generation of HCl at room temperature from a mixture of sodium chloride and glycine was confirmed through spectrophotometric monitoring of the pH. Hydrolytic and chlorination reactions were demonstrated through monitoring of formation of HMF and chlorinated products under pyrolytic conditions using glucose or sucrose and amino acid mixtures.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
2,3-Dichloro-1-propanol, ≥97.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
SAFC
L-Threonine
Supelco
Styrene, analytical standard
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Glycidol, 96%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Proline, European Pharmacopoeia (EP) Reference Standard
L-Threonine, European Pharmacopoeia (EP) Reference Standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-Glycidol, analytical standard
Supelco
Glycerol, analytical standard
Supelco
Furfuryl alcohol, analytical standard
Levodropropizine impurity C, European Pharmacopoeia (EP) Reference Standard
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%