Merck
  • Paclitaxel/epigallocatechin gallate coloaded liposome: a synergistic delivery to control the invasiveness of MDA-MB-231 breast cancer cells.
Paclitaxel/epigallocatechin gallate coloaded liposome: a synergistic delivery to control the invasiveness of MDA-MB-231 breast cancer cells.
Colloids and surfaces. B, Biointerfaces (2014-12-02)
Satiesh Kumar Ramadass, Niranjana Vaighya Anantharaman, Saravanan Subramanian, Srinivasan Sivasubramanian, Balaraman Madhan
ABSTRACT
Matrix metalloproteinases (MMPs) have been investigated as a potential target for treating invasive breast cancers. The chemotherapy for breast cancer is often prescribed as a combination of drugs. The present study investigates a novel strategy of combining a MMP inhibitor, Epigallocatechin gallate (EGCG), along with an anticancer drug, Paclitaxel (PTX), in the form of a liposomal co-delivery system. The developed PTX/EGCG co-loaded liposomes showed an entrapment of 77.11±2.30% and 59.11±3.51% for PTX and EGCG, respectively. The in vitro efficacy of the liposomes was assessed by their ability to promote apoptosis and curtail cell invasion. On all parameters, namely cytotoxicity and caspase-3 activity that are indicators of apoptosis, and MMP-2 and - 9 inhibition and invasion assays that are indicators of cell invasion, the PTX/EGCG co-loaded liposomes showed better results than each of the individual drug loaded liposomes. These findings demonstrate the synergistic outcome of PTX/EGCG combination and indicate the suitability of PTX/EGCG co-loaded liposomes for the treatment of invasive breast cancer.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Epigallocatechin gallate, primary reference standard
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material