Merck
  • Vagus nerve stimulation has antidepressant effects in the kainic acid model for temporal lobe epilepsy.
Vagus nerve stimulation has antidepressant effects in the kainic acid model for temporal lobe epilepsy.
Brain stimulation (2014-12-03)
Annelies Grimonprez, Robrecht Raedt, Ine Dauwe, Lies Mollet, Lars Emil Larsen, Alfred Meurs, Veerle De Herdt, Wytse Wadman, Jean Delbeke, Kristl Vonck, Paul Boon
ABSTRACT
Depression is the most common psychiatric comorbidity in epilepsy patients. The lack of success with current pharmacological interventions for this patient population, highlights the importance of optimizing non-pharmacological neuromodulatory treatments such as vagus nerve stimulation (VNS). Studies on the antidepressant effect of VNS in epilepsy patients may be confounded by concurrent anti-epileptic drug therapy. To date, studies in epilepsy models overcoming this problem are lacking. We investigated whether VNS affects anhedonia, a key symptom of major depression, in the kainic acid rat model for temporal lobe epilepsy. Anhedonia was assessed in kainic acid (KA) and saline (SAL) injected rats using the saccharin preference test (SPT). To exclude differences in taste perception, the quinine aversion test (QAT) was performed. Both groups were randomly subdivided in a VNS and a SHAM group, yielding 4 experimental arms: KA-VNS, KA-SHAM, SAL-VNS and SAL-SHAM. Both VNS groups received 2 weeks of VNS, while the SHAM groups were not stimulated. Thereafter, the SPT and QAT were repeated. Saccharin preference was significantly reduced in the KA compared to the SAL rats (P < 0.05), without differences in quinine aversion. Two weeks of VNS significantly increased the saccharin preference in the KA-VNS group (P < 0.05), while it had no effect on quinine aversion. No effects of VNS or SHAM were found in the other groups. The KA rats displayed anhedonia which was significantly decreased by VNS, indicating that this neuromodulatory treatment could likewise diminish depressive symptoms in patients suffering from temporal lobe epilepsy and comorbid depression.
Sigma-Aldrich
Quinine, 90%
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Quinine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
Saccharin, ≥98%
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Saccharin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%