Merck
  • Exploiting the buccal mucosa as an alternative route for the delivery of donepezil hydrochloride.
Exploiting the buccal mucosa as an alternative route for the delivery of donepezil hydrochloride.
Journal of pharmaceutical sciences (2014-04-02)
Thiago Caon, Yijun Pan, Cláudia M O Simões, Joseph A Nicolazzo
ABSTRACT
The potential of the buccal mucosa as an alternative route for the systemic delivery of donepezil (DPZ) hydrochloride, and the impact of various skin penetration enhancers on DPZ buccal permeability, was assessed using an in vitro model. DPZ was applied to porcine buccal mucosa in modified Ussing chambers either alone (20 μg/mL) or with different treatment protocols of various enhancers including Azone® (AZ), deoxycholic acid (DA), polyethylene glycol (PEG) 400, and oleic acid (OA)-PEG 400. DPZ permeated the buccal mucosa very rapidly with a permeability coefficient of 35.6 ± 4.9 × 10(-6) cm/s, which was not significantly affected by AZ pretreatment. Coapplication of DA 0.6% (w/w), but not DA 0.01% (w/w), reduced the buccal permeation of DPZ (3.5-fold), and PEG 400 reduced the absorption of DPZ in a dose-dependent manner (1.6- and 18.0-fold reduction at 5% and 50%, w/w, PEG 400, respectively). Coapplication of a combination of OA 1% (v/w) and PEG 400 5% (w/w) further reduced DPZ permeability (5.5-fold), which was demonstrated to result from excipient-induced DPZ precipitation as assessed by light microscopy analysis. These results confirm the feasibility of a novel buccal delivery system for Alzheimer's disease, and suggest various approaches that may be exploited for controlled buccal delivery of DPZ.
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)