Merck
  • Generation of monocyte-derived insulin-producing cells from non-human primates according to an optimized protocol for the generation of PCMO-derived insulin-producing cells.
Generation of monocyte-derived insulin-producing cells from non-human primates according to an optimized protocol for the generation of PCMO-derived insulin-producing cells.
Journal of clinical research in pediatric endocrinology (2014-06-17)
Jessica Walter, Ole Harder, Fred Faendrich, Maren Schulze
ABSTRACT
The vision of potential autologous cell therapy for the cure of diabetes encourages ongoing research. According to a previously published protocol for the generation of insulin-producing cells from human monocytes, we analyzed whether the addition of growth factors could increase insulin production. This protocol was then transferred to a non-human primate model by using either blood- or spleen-derived monocytes. Human monocytes were treated to dedifferentiate into programmable cells of monocytic origin (PCMO). In addition to the published protocol, PCMOs were then treated with either activin A, betacellulin, exendin 3 or 4. Cells were characterized by protein expression of insulin, Pdx-1, C-peptide and Glut-2. After identifying the optimal protocol, monocytes from baboon blood were isolated and the procedure was repeated. Spleen monocytes following splenectomy of a live baboon were differentiated and analyzed in the same manner and calculated in number and volume. Insulin content of human cells was highest when cells were treated with activin A and their insulin content was 13,000 µU/1 million cells. Insulin-producing cells form primate monocytes could successfully be generated despite using human growth factors and serum. Expression of insulin, Pdx-1, C-peptide and Glut-2 was comparable to that of human neo-islets. Total insulin content of activin A-treated baboon monocytes was 16,000 µU/1 million cells. We were able to show that insulin-producing cells can be generated from baboon monocytes with human growth factors. The amount generated from one spleen could be enough to cure a baboon from experimentally induced diabetes in an autologous cell transplant setting.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard