Merck
  • eNOS plays an important role in the regulation of colonic inflammation: a novel therapeutic target and a predictive marker for the prognosis of ulcerative colitis.
eNOS plays an important role in the regulation of colonic inflammation: a novel therapeutic target and a predictive marker for the prognosis of ulcerative colitis.
Free radical research (2014-10-21)
N Okaniwa, M Sasaki, T Mizushima, N Ogasawara, Y Funaki, T Joh, K Kasugai
ABSTRACT
We reported that deficiency of the eNOS protein exacerbates colitis induced by dextran sodium sulfate (DSS-induced colitis). However, the role of eNOS in colitis remains controversial. Therefore, we studied how over-expression of eNOS affected this inflammatory condition, using vascular endothelial cells and mice as in vitro and in vivo models, respectively. Furthermore, we investigated the influence of a polymorphism in the eNOS gene on the clinical features of ulcerative colitis (UC). We examined the effect of eNOS overexpression on the expression of adhesion molecules in the endothelium and assessed the degree of DSS-induced colitis in eNOS transgenic (eNOS-Tg) mice. We also investigated the relationship between a polymorphism in the eNOS gene and clinical features of patients with UC. The expression of adhesion molecules, under inflammatory conditions, was attenuated in eNOS gene-transfected vascular endothelial cells, as measured by western blot analysis. Symptoms of DSS-induced colitis were likewise attenuated in eNOS-Tg mice, which exhibited lower weight loss, mortality, histological damage (by inflammation score and crypt damage score), and colonic myeloperoxidase activity, tumor necrosis factor-α expression, and MAdCAM-1 expression than in wild-type mice. Furthermore, there was a significant relationship between intractable cases of UC and a polymorphism in the eNOS gene promoter (c.-786 T > C) that decreases eNOS expression. The eNOS gene plays an important role in the regulation of colonic inflammation. The level of eNOS expression may be a predictive marker for prognosis of UC, and eNOS expression may be a novel therapeutic target.
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture