Merck
  • Efficacy of gastro-retentive forms of ecabet sodium in the treatment of gastric ulcer in rats.
Efficacy of gastro-retentive forms of ecabet sodium in the treatment of gastric ulcer in rats.
Archives of pharmacal research (2013-11-21)
Ju-Young Kim, Hyun-Ju Bae, Jungsub Choi, Jong-Rae Lim, Sang-Wook Kim, Sung-Hoon Lee, Eun-Seok Park
ABSTRACT
The purpose of the present study is to investigate the influence of gastric retention of ecabet sodium (ECS) on its mucoprotective effect in rat ulcer models. Mini-tablets containing 9 mg ECS were prepared using the direct compression method. The release rates of ECS mini-tablets were controlled by the amount and viscosity grade of hydroxypropylmethyl cellulose incorporated. Gastric retention of ECS mini-tablets after oral administration to rats was visually confirmed using a fluorescence imaging system. Because ECS mini-tablets exhibited size-dependent gastric retention, their gastric retention time was prolonged as the release rate decreased. In the in vivo efficacy study, gastro-retentive dosage forms of ECS did not influence the mucoprotective effect in the immediate irritation model but enhanced the effect in the delayed irritation model compared with ECS suspension. This finding indicates that the duration of the mucoprotective effect of ECS can be extended by the employment of gastro-retentive dosage formulations and provides a rationale for development of ECS gastro-retentive dosage forms.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Magnesium stearate, technical grade
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Ketoprofen, VETRANAL®, analytical standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%