Merck
  • The role of G-protein-coupled receptor 120 in fatty acids sensing in chicken oral tissues.
The role of G-protein-coupled receptor 120 in fatty acids sensing in chicken oral tissues.
Biochemical and biophysical research communications (2015-02-07)
Ryo Sawamura, Yuko Kawabata, Fuminori Kawabata, Shotaro Nishimura, Shoji Tabata
ABSTRACT
Clarification of the mechanism of chickens' taste sense will provide meaningful information for creating and improving new feedstuff for chickens, because the character of taste receptors in oral tissues affects feeding behavior in animals. Although fatty acids are partly recognized via G-protein coupled receptor 120 (GPR120) for fat taste in mammalian oral tissues, the fat taste receptor of chickens has not been elucidated. Here we cloned chicken GPR120 (cGPR120) from the chicken palate, which contains taste buds. By using Ca(2+) imaging methods, we identified oleic acid and linoleic acid as cGPR120 agonists. Interestingly, in a behavioral study the chickens preferred corn oil-rich feed over mineral oil (control oil)-rich feed. Because corn oil contains high amounts of oleic acid and linoleic acid, this result was thought to be reasonable. Taken together, the present results suggest that cGPR120 is one of the functional fat taste receptors in chickens.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture