Merck
  • Proteolysis is a confounding factor in the interpretation of faecal calprotectin.
Proteolysis is a confounding factor in the interpretation of faecal calprotectin.
Clinical chemistry and laboratory medicine (2014-07-06)
Els N Dumoulin, Stephanie Van Biervliet, Michel R Langlois, Joris R Delanghe
ABSTRACT
Calprotectin is a 36 kDa calcium and zinc binding protein. An increased level of calprotectin points towards inflammatory bowel disease. However, the biomarker calprotectin shows 14 potential cleavages sites for trypsin. Next to trypsin, also the presence of its inhibitor α1-antitrypsin after a gastrointestinal bleeding may affect calprotectin testing. In this study, effects of trypsin and α1-antitrypsin as potential confounders for faecal calprotectin testing are investigated. An in vitro model was created. As calprotectin source, leukocytes were isolated and subsequently lysed (1% Triton X-100) and diluted in faecal matrix. Trypsin digestion was carried out by adding trypsin. Incubation occurred for 24 h or 48 h (37 °C). To study the influence of α1-antitrypsin on trypsin, the same experiment was repeated after adding serum containing α1-antitrypsin. In vitro experiments enabled monitoring of the faecal calprotectin digestion, leading to loss of immunoreactivity. Trypsin activity was a potential confounder in the interpretation of calprotectin, in particular for proximal lesions, where exposure of calprotectin to trypsin is prolonged. Relative calprotectin loss was proportional to the amount of trypsin. Decrease of calprotectin was more pronounced after 48 h of incubation in comparison to 24 h of incubation. Analogue experiments also showed stable calprotectin values after adding α1-antitrypsin. Transit time, trypsin activity and addition of blood as a source of α1-antitrypsin may be regarded as potential confounders in the interpretation of calprotectin results. Age-related cut-off values depending on the anatomical localisation of the lesions could improve the diagnostic efficiency of calprotectin testing.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)