Merck
  • Enhanced biosensing resolution with foundry fabricated individually addressable dual-gated ISFETs.
Enhanced biosensing resolution with foundry fabricated individually addressable dual-gated ISFETs.
Analytical chemistry (2014-07-30)
Carlos Duarte-Guevara, Fei-Lung Lai, Chun-Wen Cheng, Bobby Reddy, Eric Salm, Vikhram Swaminathan, Ying-Kit Tsui, Hsiao Chin Tuan, Alex Kalnitsky, Yi-Shao Liu, Rashid Bashir
ABSTRACT
The adaptation of semiconductor technologies for biological applications may lead to a new era of inexpensive, sensitive, and portable diagnostics. At the core of these developing technologies is the ion-sensitive field-effect transistor (ISFET), a biochemical to electrical transducer with seamless integration to electronic systems. We present a novel structure for a true dual-gated ISFET that is fabricated with a silicon-on-insulator (SOI) complementary metal-oxide-semiconductor process by Taiwan Semiconductor Manufacturing Company (TSMC). In contrast to conventional SOI ISFETs, each transistor has an individually addressable back-gate and a gate oxide that is directly exposed to the solution. The elimination of the commonly used floating gate architecture reduces the chance of electrostatic discharge and increases the potential achievable transistor density. We show that when operated in a "dual-gate" mode, the transistor response can exhibit sensitivities to pH changes beyond the Nernst limit. This enhancement in sensitivity was shown to increase the sensor's signal-to-noise ratio, allowing the device to resolve smaller pH changes. An improved resolution can be used to enhance small signals and increase the sensor accuracy when monitoring small pH dynamics in biological reactions. As a proof of concept, we demonstrate that the amplified sensitivity and improved resolution result in a shorter detection time and a larger output signal of a loop-mediated isothermal DNA amplification reaction (LAMP) targeting a pathogenic bacteria gene, showing benefits of the new structure for biosensing applications.
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%